ENA448967986 Enamine

1-acetyl-3-(3-amino-2-methylphenyl)imidazolidin-2-one

  • MDL number MFCD28505679
  • Cas Number : 1803606-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201216
Availability details for
Quantity
  • Availability
  • Add to Cart