ENA448968002 Enamine

1-(pyridin-2-yl)cyclobutane-1-carbonitrile

  • MDL number MFCD16622259
  • Cas Number : 485828-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202731
Availability details for
Quantity
  • Availability
  • Add to Cart