ENA448968007 Enamine

4-methyl-7-(methylamino)-2H-chromen-2-one

  • MDL number MFCD00489511
  • Cas Number : 40568-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203019
Availability details for
Quantity
  • Availability
  • Add to Cart