ENA448968017 Enamine

ethyl 3-bromo-2,2-dimethylpropanoate

  • MDL number MFCD24448073
  • Cas Number : 2843-18-7
  • Molecular Weight : 209.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-bromo-2,2-dimethylpropanoate
Inchi Code 1S/C7H13BrO2/c1-4-10-6(9)7(2,3)5-8/h4-5H2,1-3H3
Inchi Key OKSCUZSVZULTMP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-112729

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-112729
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart