ENA448968018 Enamine

2-(1-methylcyclopentyl)acetonitrile

  • MDL number MFCD28505691
  • Cas Number : 1803591-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204122
Availability details for
Quantity
  • Availability
  • Add to Cart