ENA448968021 Enamine

2,4-dichloro-6-methylpyridine-3-carbonitrile

  • MDL number MFCD11847537
  • Cas Number : 38367-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204217
Availability details for
Quantity
  • Availability
  • Add to Cart