ENA448968032 Enamine

1-benzyl-1H-1,2,3-triazole-4-carbonitrile

  • MDL number MFCD00826321
  • Cas Number : 202003-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204811
Availability details for
Quantity
  • Availability
  • Add to Cart