ENA448968033 Enamine

4-(difluoromethoxy)quinoline

  • MDL number MFCD18416201
  • Cas Number : 1261458-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204832
Availability details for
Quantity
  • Availability
  • Add to Cart