ENA448968034 Enamine

2-amino-1-[3-(trifluoromethoxy)phenyl]ethan-1-ol

  • MDL number MFCD21662309
  • Cas Number : 852392-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114495
Availability details for
Quantity
  • Availability
  • Add to Cart