ENA448968041 Enamine

2-(1-methyl-4-sulfamoyl-1H-pyrrol-2-yl)acetic acid

  • MDL number MFCD28505699
  • Cas Number : 1803592-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205443
Availability details for
Quantity
  • Availability
  • Add to Cart