ENA448968055 Enamine

N-[1-(2-phenyl-1,3-thiazol-4-yl)ethylidene]hydroxylamine

  • Cas Number : 57372-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206135
Availability details for
Quantity
  • Availability
  • Add to Cart