ENA448968070 Enamine

5-bromo-4-chloro-2-methylpyridin-1-ium-1-olate

  • MDL number MFCD25954706
  • Cas Number : 103971-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206703
Availability details for
Quantity
  • Availability
  • Add to Cart