ENA448968081 Enamine

1-(2-nitrophenyl)-1H-1,2,4-triazole-3-carbonitrile

  • MDL number MFCD17241747
  • Cas Number : 1250607-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207016
Availability details for
Quantity
  • Availability
  • Add to Cart