ENA448968082 Enamine

N-(4-methylpyrimidin-2-yl)-4-nitrobenzene-1-sulfonamide

  • MDL number MFCD02930745
  • Cas Number : 439928-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207037
Availability details for
Quantity
  • Availability
  • Add to Cart