ENA448968088 Enamine

5,6-dibromopyridine-3-sulfonyl chloride

  • MDL number MFCD18411511
  • Cas Number : 1261883-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207269
Availability details for
Quantity
  • Availability
  • Add to Cart