ENA448968102 Enamine

1-(3-chloro-4-ethylphenyl)piperazine hydrochloride

  • MDL number MFCD28505718
  • Cas Number : 1803588-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207823
Availability details for
Quantity
  • Availability
  • Add to Cart