ENA448968104 Enamine

2-(methylamino)-3-nitropyridine-4-carboxylic acid

  • MDL number MFCD28505719
  • Cas Number : 1803565-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207887
Availability details for
Quantity
  • Availability
  • Add to Cart