ENA448968106 Enamine

1-(benzyloxy)-2-nitro-4-(trifluoromethyl)benzene

  • MDL number MFCD01422843
  • Cas Number : 135328-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207989
Availability details for
Quantity
  • Availability
  • Add to Cart