ENA448968110 Enamine

1-chloro-5-methoxy-4-methyl-2-nitrobenzene

  • MDL number MFCD28364383
  • Cas Number : 97655-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208098
Availability details for
Quantity
  • Availability
  • Add to Cart