ENA448968114 Enamine

2H,3H-1lambda6-[1,2]thiazolo[5,4-c]pyridine-1,1,3-trione

  • MDL number MFCD08695605
  • Cas Number : 142141-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208197
Availability details for
Quantity
  • Availability
  • Add to Cart