ENA448968131 Enamine

6,6-dimethylheptanoic acid

  • MDL number MFCD01311828
  • Cas Number : 15898-92-7
  • Molecular Weight : 158.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-208555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208555
Availability details for
Quantity
  • Availability
  • Add to Cart