ENA448968174 Enamine

[2-(methoxymethyl)phenyl]methanesulfonyl chloride

  • MDL number MFCD21267569
  • Cas Number : 1496892-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134492
Availability details for
Quantity
  • Availability
  • Add to Cart