ENA448968192 Enamine

3-fluoro-4-(piperidin-4-yloxy)benzonitrile hydrochloride

  • MDL number MFCD28505751
  • Cas Number : 1799968-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210106
Availability details for
Quantity
  • Availability
  • Add to Cart