ENA448968194 Enamine

2,5-difluoro-4-(morpholin-4-yl)aniline

  • MDL number MFCD28118182
  • Cas Number : 1238704-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210119
Availability details for
Quantity
  • Availability
  • Add to Cart