ENA448968211 Enamine

5-amino-3-phenyl-1,2-oxazole-4-carbonitrile

  • MDL number MFCD09703133
  • Cas Number : 14246-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-192
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136546
Availability details for
Quantity
  • Availability
  • Add to Cart