ENA448968212 Enamine

ethyl N-[(4-chloropyridin-2-yl)carbamothioyl]carbamate

  • MDL number MFCD23131139
  • Cas Number : 1131410-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210697
Availability details for
Quantity
  • Availability
  • Add to Cart