ENA448968213 Enamine

5-(4-bromophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02690769
  • Cas Number : 73040-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210699
Availability details for
Quantity
  • Availability
  • Add to Cart