ENA448968228 Enamine

methyl 1-(bromomethyl)-2-oxocycloheptane-1-carboxylate

  • MDL number MFCD28505760
  • Cas Number : 107987-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210914
Availability details for
Quantity
  • Availability
  • Add to Cart