ENA448968240 Enamine

2-bromoprop-2-enal

  • MDL number MFCD00061520
  • Cas Number : 14925-39-4
  • Molecular Weight : 134.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 97
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100680

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P273,P280,P301+P310,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P311,P312,P330,P361,P363,P391,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-100680
pictograms GHS05,GHS06,GHS09
hstatements H300,H311,H314,H331,H400
Availability details for
Quantity
  • Availability
  • Add to Cart