ENA448968244 Enamine

4-ethoxypyridine-3-sulfonamide

  • MDL number MFCD28505769
  • Cas Number : 1229666-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211042
Availability details for
Quantity
  • Availability
  • Add to Cart