ENA448968246 Enamine

5-cyanofuran-2-sulfonyl chloride

  • MDL number MFCD19201179
  • Cas Number : 98027-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211053
Availability details for
Quantity
  • Availability
  • Add to Cart