ENA448968250 Enamine

5,6-dichloro-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD22398618
  • Cas Number : 1781138-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147291
Availability details for
Quantity
  • Availability
  • Add to Cart