ENA448968251 Enamine

methyl (2R)-2-{[(2S)-piperidin-2-yl]formamido}propanoate hydrochloride

  • MDL number MFCD28505774
  • Cas Number : 1807937-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211097
Availability details for
Quantity
  • Availability
  • Add to Cart