ENA448968259 Enamine

6H,7H,8H,9H,11H-pyrido[2,1-b]quinazolin-11-one

  • MDL number MFCD00661747
  • Cas Number : 2446-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211116
Availability details for
Quantity
  • Availability
  • Add to Cart