ENA448968268 Enamine

1-(propan-2-yl)-3-(propylamino)pyrrolidine-2,5-dione

  • MDL number MFCD12098633
  • Cas Number : 1218600-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211162
Availability details for
Quantity
  • Availability
  • Add to Cart