ENA448968278 Enamine

6-bromo-2,1-benzoxazole

  • MDL number MFCD16995918
  • Cas Number : 139557-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211219
Availability details for
Quantity
  • Availability
  • Add to Cart