ENA448968310 Enamine

5-methanesulfonyl-2-(2-methoxyethoxy)aniline

  • MDL number MFCD12172818
  • Cas Number : 1154235-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211367
Availability details for
Quantity
  • Availability
  • Add to Cart