ENA448968313 Enamine

2-fluoro-1-methoxy-3-nitrobenzene

  • MDL number MFCD13185616
  • Cas Number : 445-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211392

Safety Information

signalword Danger
pcodes P264,P270,P273,P301+P310,P330,P405,P501
hcodes T
MSDS https://www.enaminestore.com/catalog/EN300-211392
pictograms GHS06
hstatements H301,H412
Availability details for
Quantity
  • Availability
  • Add to Cart