ENA448968317 Enamine

[2-(methylamino)ethyl][2-(pyrrolidin-1-yl)ethyl]amine trihydrochloride

  • MDL number MFCD28505800
  • Cas Number : 1803606-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211400
Availability details for
Quantity
  • Availability
  • Add to Cart