ENA448968324 Enamine

2-bromo-3-phenylpyrazine

  • MDL number MFCD18447738
  • Cas Number : 2882-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211410
Availability details for
Quantity
  • Availability
  • Add to Cart