ENA448968329 Enamine

3-amino-N-(5-ethyl-1,3,4-thiadiazol-2-yl)-2-methylbutanamide hydrochloride

  • MDL number MFCD28505805
  • Cas Number : 1803589-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211497
Availability details for
Quantity
  • Availability
  • Add to Cart