ENA448968333 Enamine

N-(2-aminoethyl)-2-bromo-5-methylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD28505809
  • Cas Number : 1604485-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211502
Availability details for
Quantity
  • Availability
  • Add to Cart