ENA448968337 Enamine

4-(1H-1,2,3-triazol-1-yl)pyridin-3-amine

  • MDL number MFCD28505813
  • Cas Number : 1803562-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211507
Availability details for
Quantity
  • Availability
  • Add to Cart