ENA448968338 Enamine

N-(2-methylpropyl)aniline hydrochloride

  • MDL number MFCD28505814
  • Cas Number : 31601-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-195
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211508
Availability details for
Quantity
  • Availability
  • Add to Cart