ENA448968352 Enamine

3-bromo-5-methyl-1,2-oxazole

  • MDL number MFCD06660140
  • Cas Number : 25741-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170702

Safety Information

signalword Warning
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-170702
pictograms GHS07
hstatements H227,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart