ENA448968361 Enamine

1-(2-chlorophenyl)-1,4-diazepane dihydrochloride

  • MDL number MFCD28505825
  • Cas Number : 1803612-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211605
Availability details for
Quantity
  • Availability
  • Add to Cart