ENA448968366 Enamine

5-(1,4,5,6-tetrahydropyrimidin-2-yl)pyridin-2-amine

  • MDL number MFCD07776882
  • Cas Number : 868260-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211611
Availability details for
Quantity
  • Availability
  • Add to Cart