ENA448968371 Enamine

4-(2-hydroxyethoxy)oxane-4-carbonitrile

  • MDL number MFCD28151985
  • Cas Number : 1701845-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170833
Availability details for
Quantity
  • Availability
  • Add to Cart