ENA448968378 Enamine

methyl 3-(aminomethyl)pyrrolidine-1-carboxylate hydrochloride

  • MDL number MFCD28505830
  • Cas Number : 1803612-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211655
Availability details for
Quantity
  • Availability
  • Add to Cart