ENA448968380 Enamine

6-(phenylamino)pyrimidine-4-carboxylic acid

  • MDL number MFCD21884845
  • Cas Number : 1208980-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211657
Availability details for
Quantity
  • Availability
  • Add to Cart